missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF25 Monoclonal Antibody (OTI2B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505428
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the kinesin-like protein family. Protein family members are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. However, the particular function of this gene product has not yet been determined. Two alternatively spliced transcript variants which encode products have been described. Other splice variants have been found that lack exon 2 and the initiation codon for translation.Specifications
| KIF25 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UIL4 | |
| KIF25 | |
| Full length human recombit protein of human KIF25 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2B4 | |
| Unconjugated | |
| KIF25 | |
| KNSL3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3834 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction