Learn More
KIF25 Monoclonal Antibody (OTI2C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505426
Encompass_Preferred
 View more versions of this product
Product Code. 17583862
  • € 540.87 / 100µL
Description
The protein encoded by this gene is a member of the kinesin-like protein family. Protein family members are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. However, the particular function of this gene product has not yet been determined. Two alternatively spliced transcript variants which encode products have been described. Other splice variants have been found that lack exon 2 and the initiation codon for translation.
Specifications
Specifications
KIF25
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UIL4
KIF25
Full length human recombit protein of human KIF25 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2C9
Unconjugated
KIF25
KNSL3
Mouse
Affinity Chromatography
RUO
3834
-20° C, Avoid Freeze/Thaw Cycles