missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF26B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17422-1-AP-20UL
This item is not returnable.
View return policy
Description
KIF26B is a member of kinesin-11 family and plays important role in embryogenesis. It is a downstream target of Sall1 and is essential for kidney development. KIF26B is also involved in tumorigenesis.Specifications
| KIF26B | |
| Polyclonal | |
| Unconjugated | |
| KIF26B | |
| KIF26B, kinesin family member 26B, Kinesin like protein KIF26B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269152, 305012, 55083 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q2KJY2, Q7TNC6 | |
| Kif26b | |
| KIF26B Fusion Protein Ag11446 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction