missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIF2A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110684

Product Code. 17198603

  • € 630.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DNESVTVEWI ENGDTKGKEI DLESIFSLNP DLVPDEEIEP SPETPPPPAS SAKVNKIVKN RRTVASIKND PPSRDNRVVG SARARPSQFP EQSSSAQQNG SVSDISPVQA Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 92%.

KIF2A, a gene encoding the kinesin family member 2A protein, plays a pivotal role in neuronal development and function. This gene is part of the kinesin-13 subfamily, characterized by its unique ability to depolymerize microtubules, which are essential components of the cytoskeleton responsible for maintaining cell shape, enabling motility, and facilitating intracellular transport. KIF2A is particularly significant in neural settings, where it regulates microtubule dynamics crucial for neuronal migration and dendritic development. Dysfunctional KIF2A, due to mutations, has been linked to early-onset neurodegenerative diseases such as progressive microcephaly and mental disabilities due to impaired microtubule regulation, reflecting its foundational role in correct brain development and architecture. Additionally, KIF2A is involved in spindle assembly and chromosome segregation during cell division, further cementing its importance in both neurodevelopment and general cellular functions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KIF2A
Polyclonal
Unconjugated
Kif2a
C530030B14Rik; CDCBM3; HK2; hypothetical protein; KIF2; kif2a; kif2-a; kif2a {ECO:0000250; kif2a.L; kinesin family member 2A; kinesin heavy chain family, member 2; kinesin heavy chain member 2; kinesin heavy chain member 2A; kinesin heavy chain member 2A L homeolog; Kinesin, heavy chain, 2; kinesin-2; Kinesin-like protein KIF2A; kinesin-related protein XKIF2; Kns2; M-kinesin; RCJMB04_9a7; UniProtKB:O00139}; Unknown (protein for MGC:133806); XELAEV_18008243mg
Rabbit
Antigen affinity chromatography
RUO
3796, 84391
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O00139, Q9WV63
Kif2a
Recombinant Protein corresonding to Human KIF2A. Recombinant protein control fragment (Product #RP-102374).
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.