missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF2C Antibody (KIF2C/4707), Novus Biologicals™
Mouse Monoclonal Antibody
€ 249.00 - € 494.00
Specifications
| Antigen | KIF2C |
|---|---|
| Clone | KIF2C/4707 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620285
|
Novus Biologicals
NBP3-20554-100ug |
100 μg |
€ 494.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654445
|
Novus Biologicals
NBP3-20554-20ug |
20 μg |
€ 249.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| KIF2C | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cellular Markers, DNA Repair | |
| 10mM PBS with 0.05% BSA | |
| 11004 | |
| IgG κ | |
| Protein A or G purified |
| KIF2C/4707 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin | |
| Recombinant fragment (around aa500-700) of human KIF2C protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title