missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF2C Antibody (KIF2C/6523), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20670-20ug
This item is not returnable.
View return policy
Description
KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| KIF2C | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin | |
| Recombinant fragment (around aa500-700) of human KIF2C protein (exact sequence is proprietary) | |
| 20 μg | |
| Cancer, Cell Biology, Cellular Markers, DNA Repair | |
| 11004 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin) | |
| KIF2C/6523 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction