Learn More
Invitrogen™ KIF4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587703
Description
Immunogen sequence: LIGELVSSKI QVSKLESSLK QSKTSCADMQ KMLFEERNHF AEIETELQAE LVRMEQQHQE KVLYLLSQLQ QSQMAEKQLE ESVSEKEQQL LSTLKCQDEE LEKMREVCEQ NQQLLRENEI IKQKLTLLQV ASRQKHLPKD TLLSPDSSFE YVPPKPKPSR VKEKFLEQSM DIEDLKYCSE HSVNEHEDGD GDDDEGDDEE WKPTKLVKVS R; Positive Samples: U-87MG, LO2, 293T, SW480; Cellular Location: Chromosome, Cytoplasm, Midbody, Nucleus matrix, cytoskeleton, spindle.
This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
Specifications
| KIF4A | |
| Polyclonal | |
| Unconjugated | |
| Kif4a | |
| AI323435; Chromokinesin; chromokinesin-A; Chromosome-associated kinesin KIF4; chromosome-associated kinesin KIF4A; D330050K22Rik; FLJ12530; FLJ12655; FLJ14204; FLJ20631; HSA271784; KIF4; KIF4A; KIF4G1; KIF4-G1; kinesin family member 4; kinesin family member 4A; kinesin heavy chain member 4; Kns4; MRX100; N-5 kinesin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16571, 24137, 84393 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95239, P33174 | |
| Kif4, Kif4a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 870-1080 of human KIF4A (NP_0364423). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.