missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF7 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46838AF488
This item is not returnable.
View return policy
Description
KIF7 Polyclonal antibody specifically detects KIF7 in Human samples. It is validated for Western Blot
Specifications
| KIF7 | |
| Polyclonal | |
| Western Blot | |
| EQYK340, kinesin family member 7, kinesin-like protein KIF7, MGC120653, MGC138476, MGC138478, UNQ340 | |
| The immunogen for this product maps to a region between residue 1225 and 1275 of human kinesin family member 7 using the numbering given in entry NP_940927.2 (GeneID 374654). | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells | |
| 374654 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction