missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF9 Antibody (4E9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00064147-M07
This item is not returnable.
View return policy
Description
KIF9 Monoclonal antibody specifically detects KIF9 in Human samples. It is validated for Western Blot, ELISA
Specifications
| KIF9 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| kinesin family member 9, kinesin protein 9, kinesin-like protein KIF9, MGC104186 | |
| KIF9 (NP_878905.1, 691 a.a. ∽ 789 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VDQCRHRLLMEFDIWYNESFVIPEDMQMALKPGGSIRPGMVPVNRIVSLGEDDQDKFSQLQQRVLPEGPDSISFYNAKVKIEQKHNYLKTMMGLQQAHR | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4E9 | |
| Western Blot, ELISA | |
| NP_878905 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 64147 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction