missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF9 Antibody (OTI1A2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44807
This item is not returnable.
View return policy
Description
KIF9 Monoclonal antibody specifically detects KIF9 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| KIF9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 64147 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1A2 | |
| Western Blot 1:2000 | |
| kinesin family member 9, kinesin protein 9, kinesin-like protein KIF9, MGC104186 | |
| Human recombinant protein fragment corresponding to amino acids 430-790 of human KIF9 (NP_878905) produced in E.coli. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction