Learn More
Invitrogen™ KIF9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113586
Description
Immunogen sequence: MGTRKKVHAF VRVKPTDDFA HEMIRYGDDK RSIDIHLKKD IRRGVVNNQQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 100%.
KIF9 is a protein coding gene.
Specifications
| KIF9 | |
| Polyclonal | |
| Unconjugated | |
| KIF9 | |
| KIF9; kinesin 9; kinesin family member 9; kinesin protein 9; kinesin-like protein KIF9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64147 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HAQ2 | |
| KIF9 | |
| Synthetic peptide directed towards the n terminal region of human KIF9 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.