missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFC1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-40844FR
This item is not returnable.
View return policy
Description
KIFC1 Polyclonal antibody specifically detects KIFC1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP), KnockDown
Specifications
| KIFC1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP), Knockdown Validated | |
| HSETMGC1202, kinesin family member C1, kinesin-like 2, Kinesin-like protein 2, kinesin-like protein KIFC1, Kinesin-related protein HSET, KNSL2MGC149736, MGC149737 | |
| The immunogen recognized by this antibody maps to a region between residue 625 and 673 of human Kinesin Family Member C1 (Mitotic Kinesin-Like Protein 2) using the numbering given in entry NP_002254.1 (GeneID 3833). | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 3833 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP Assay, KnockDown | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction