Learn More
DWK Life Sciences Kimble™ KIMAX™ Media Bottles — Ungraduated
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Closure Size:
33-430 mm
Unit Size:
Pack of 24
Pack of 48
Description
- Available with (Cat. No. 06-421-7) or without graduations
- 1000mL size features finger grips molded into the side wall and is available with two GPI thread sizes
- All bottles have screw-thread finish
- Without closures
Specifications
Specifications
| Capacity | 4.22 oz., 125 mL |
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4.22 oz. |
| Color | White |
| Diameter (English) | 2.1 in. |
| Material | Borosilicate Glass |
| Height (Metric) | 119 mm |
| Height (English) | 4.6 in. |
| Closure Size | 33-430 mm |
| Diameter (Metric) | 55 mm |