missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Black Phenolic Unlined Screw Thread Closures with Open Tops

Product Code. 12804871
Click to view available options
Neck Finish:
15-425
18-400
24-400
Unit Size:
Pack of 144
This item is not returnable. View return policy

Product Code. 12804871

Brand: DWK Life Sciences 7380424400

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Open top closures providing excellent chemical resistance

  • Phenolic
  • Open top
  • Autoclavable
  • Unlined
  • Black

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 24-400
For Use With (Equipment) Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.