missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Black Phenolic Unlined Screw Thread Closures with Open Tops
Click to view available options
Neck Finish:
15-425
18-400
24-400
Unit Size:
Pack of 144
Description
Open top closures providing excellent chemical resistance
- Phenolic
- Open top
- Autoclavable
- Unlined
- Black
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 144 Cs. |
| Neck Finish | 18-400 |
| Product Type | Open Top Screw Cap |
| For Use With (Equipment) | Bottle |
| Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction