Learn More
DWK Life Sciences Kimble™ KIMAX™ Baffled Culture Flasks
For fermentation, cell culture growth and cultivation, and genetic engineering procedures
Brand:  DWK Life Sciences 25630-2000
Additional Details : Weight : 0.55800kg
Product Code. 11750734
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
For fermentation, cell culture growth and cultivation, and genetic engineering procedures
  • Bottom baffles provide increased mixing, aeration, and gas exchange
  • Neck O.D.: 25mm (1 in.) for 125mL size; all others are Neck O.D.: 38mm (11/2 in.)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers