Learn More
DWK Life Sciences Kimble™ KIMAX™ Flasks with Quick-Release Hose Connector
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
Brand:  DWK Life Sciences 270701000
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11737989
  • € 132.00 / Each
Estimated Shipment
23-06-2021
Add to basket
Description
Description
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
  • Tubing remains permanently attached to hose barb and opposite end attaches to flask tubulation with positive threaded seal
  • Proper positioning of connector provides greater flask stability as tubing angled downward has less tendency to tilt flask
  • Graduated with approximate volume scale in durable white ceramic enamel
  • Tubing and stoppers sold separately
Specifications
Specifications
8
1000 mL
300 to 1000 mL
23.8 cm
13.5 cm
Borosilicate Glass
50 mL