Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Volumetric Unserialized Class A Pipets
Product Code. 10067642
Click to view available options
Volume (Metric):
3 mL
Length (English):
13.77 in.
Length (Metric):
35 cm
Unit Size:
Pack of 6
Product Code. 10067642
Brand: DWK Life Sciences 370073
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Kimax-51 glass, with tempered tip and top-end contouring.
  • Calibrated to contain (bottom line) and to deliver (top line) to Class A tolerances
  • Color-coded (ASTM E1273) for ease in sorting and selecting the correct pipet
  • Large numerals on the bulb indicate capacity
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Contain, To Deliver
Certifications/Compliance ASTM E969
Class A
Material Borosilicate Glass
Length (English) 13.77 in.
Tolerance ±0.01 mL
Volume (English) 0.1 oz.
Color Black
Volume (Metric) 3 mL
Length (Metric) 35 cm
Type Unserialized Volumetric Pipet
Disposable Reusable