Learn More
DWK Life Sciences Kimble™ KIMAX™ Storage/Media Bottles
Product Code. 12633225
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
Graduations:
100 to 400mL
100 to 900mL
1000 to 4000mL
20 to 80mL
2000 to 8000mL
400 to 1800mL
50 to 200mL
Unit Size:
Each
Pack of 4
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations unitSize
12633225 5000 mL 1000 to 4000mL Each
12663215 100 mL 20 to 80mL Pack of 4
12683215 250 mL 50 to 200mL Pack of 4
12693215 500 mL 100 to 400mL Pack of 4
12603225 1000 mL 100 to 900mL Pack of 4
12613225 2000 mL 400 to 1800mL Pack of 4
12623225 10,000 mL 2000 to 8000mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12633225 Supplier DWK Life Sciences Supplier No. KC143955000
Only null left
Add to basket
Add to basket
With linerless blue polypropylene caps
  • KimCote™ plastic coating helps prevent and contain spills in the event of glassware breakage
  • GL 45 screw thread bottles with white graduations and marking spots
  • Wide opening for easy access (approx. 30mm I.D.)
  • Linerless cap is blue polypropylene and autoclavable (140°C max)
  • Replaceable pour ring included
  • Ultra-clear for improved sample viewing
  • Non-slip surface — wet or dry
Applications:
Pressurized glass, chemical storage, transportation of samples, handling of hazardous chemicals, harmful biologicals and acids
TRUSTED_SUSTAINABILITY
Specifications
Capacity 169.07 oz., 5000 mL
Capacity (Metric) 5000 mL
Capacity (English) 169.07 oz.
Color Ultra Clear
Diameter (English) 7.1 in.
Diameter (English) Inner Neck 1.18 in.
Diameter (Metric) Inner Neck 30 mm
Includes Clear drip-free Polypropylene pouring ring
Material Plastic
Closure Color Blue
Graduations 1000 to 4000mL
Closure Included Yes
Graduated Yes
Mouth Wide
Height (Metric) 330 mm
Product Type Media Bottle
Height (English) 12.9 in.
Closure Material Polypropylene
Closure Size GL 45
Diameter (Metric) 181 mm
Closure Type Screw Cap