missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Storage/Media Bottles

Catalog Number 12663215
Click to view available options
Capacity:
169.07 oz., 5000 mL
17 oz., 500 mL
3.3 oz., 100 mL
33.8 oz., 1000 mL
338.1 oz., 10000mL
67.62 oz., 2000 mL
8.45 oz., 250 mL
Diameter (English):
2.2 in.
2.7 in.
3.3 in.
3.9 in.
5.3 in.
7.1 in.
8.9 in.
Graduations:
100 to 400mL
100 to 900mL
1000 to 4000mL
20 to 80mL
2000 to 8000mL
400 to 1800mL
50 to 200mL
Height (English):
3.9 in.
5.4 in.
6.9 in.
8.8 in.
10.23 in.
12.9 in.
16.1 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

With linerless blue polypropylene caps

  • KimCote™ plastic coating helps prevent and contain spills in the event of glassware breakage
  • GL 45 screw thread bottles with white graduations and marking spots
  • Wide opening for easy access (approx. 30mm I.D.)
  • Linerless cap is blue polypropylene and autoclavable (140°C max)
  • Replaceable pour ring included
  • Ultra-clear for improved sample viewing
  • Non-slip surface — wet or dry

Applications:

Pressurized glass, chemical storage, transportation of samples, handling of hazardous chemicals, harmful biologicals and acids

Capacity 3.3 oz., 100 mL
Closure Material Polypropylene
Closure Size GL 45
Closure Type Screw Cap
Diameter (Metric) 56 mm
Height (English) 3.9 in.
Height (Metric) 100 mm
Capacity (Metric) 100 mL
Capacity (English) 3.3 oz.
Color Ultra Clear
Diameter (English) 2.2 in.
Diameter (English) Inner Neck 1.18 in.
Diameter (Metric) Inner Neck 30 mm
Includes Clear drip-free Polypropylene pouring ring
Material Plastic
Closure Color Blue
Graduations 20 to 80mL
Graduated Yes
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'