Learn More
DWK Life Sciences Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
Product Code. 11827423
Click to view available options
Length (Metric):
300 mm
400 mm
500 mm
600 mm
Diameter (Metric) Inner:
11 mm
19 mm
22 mm
25 mm
41 mm
Column Capacity:
20 mL
110 mL
250 mL
790 mL
Includes:
Fritted disc
Without fritted disc
Unit Size:
Each
Product Code. 11827423
Brand: DWK Life Sciences 4203200245
Only null left
Add to basket
Add to basket
With or without fritted disc
  • Borosilicate glass
  • Sealed-in coarse porosity (40 to 60μm) fritted disc supports packing in K420320 Series columns
  • Use glass wool to support packing in K420300 Series columns (packing not included)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Many separations in adsorption and ion exchange chromatography
Diameter (Metric) Inner 25 mm
Type Adsorption/Ion Exchange Chromatography Column
Pore Size 40 to 60 μm
Column Capacity 250 mL
Length (Metric) 500 mm
Material Borosilicate Glass
Includes Fritted disc