Learn More
DWK Life Sciences Kimble™ Kontes™ All-Glass Potter-Elvehjem Tissue Grinders
Designed for motor drive using 788000 Stirrer Adapter
Brand: DWK Life Sciences 885500-0021
Additional Details : Weight : 1.00000kg
Description
- All-glass construction
- Clearance between pestles and tubes is 0.004 to 0.006 in.
- Replacement components are available and completely interchangeable
Specifications
21 | |
0.00015 to 0.0002mm | |
6mm | |
5 mL | |
Tissue Grinder Tube | |
All Glass Potter-Elvehjem tissue grinders | |
205mm | |
140mm |
0.004 to 0.006 in. | |
0.23 in. | |
0.16 oz. | |
Ground Glass | |
Designed for motor drive using 788000 stirrer adapter | |
8.07 in. | |
5.5 in. | |
0.16 oz., 5mL |