Learn More
DWK Life Sciences Kimble™ Kontes™ Chromaflex™ Columns
Product Code. 10708995
Change view
Click to view available options
Length (Metric):
150 mm
300 mm
1200 mm
Diameter (Metric) Inner:
10 mm
25 mm
48 mm
Column Capacity:
24 mL
147 mL
271 mL
2172 mL
Max. Pressure:
100 psi
49.31 psi
75.41 psi
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Column Capacity Max. Pressure unitSize
10708995 1200 mm 48 mm 2172 mL 49.31 psi Each
11853193 150 mm 48 mm 271 mL 49.31 psi Each
12716788 300 mm 25 mm 147 mL 75.41 psi Each
10103952 300 mm 10 mm 24 mL 100 psi Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10708995 Supplier DWK Life Sciences Supplier No. 4208301220
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Includes
Glass barrel, Two PTFE end fittings with 20μm porosity polyethylene bed supports, 5 ft. (1.5m) of PTFE tubing (1/16 in. O.D. tubing with 1.0cm I.D. columns, 1/8 in. O.D. tubing with 2.5 and 4.8cm I.D. columns), Two flangeless tubing nuts, Two flangeless ferrules
Ideal for medium-pressure chromatography with aqueous mobile phases
  • Threaded
  • Feature PTFE-shielded O-rings and end fittings
  • PTFE layer is curled over the O-ring to form a solvent- and leak-resistant seal
  • Solvents contact only PTFE and borosilicate glass surfaces
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Medium pressure chromatography with aqueous mobile phases
Temperature (Metric) Range 0 to 50°C
Diameter (Metric) Inner 48 mm
Type Chromatography Column
Max. Pressure 49.31 psi
Pore Size 20 μm
Column Capacity 2172 mL
Length (Metric) 1200 mm
Material Borosilicate Glass Type I Class A
Support Material HDPE
Connector Types 1/4-28 Flangeless fitting
Includes Two PTFE end fittings with 20um porosity Polyethylene bed supports, 5 ft. PTFE tubing, 2 flangeless tubing nuts and 2 flangeless ferrules
Sterilization Method Ethylene Oxide, 2N NaOH or 100% Ethanol