Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Pestle For All-Glass Dounce Tissue Grinders
Click to view available options
Capacity (Metric):
15 mL
40 mL
Product Type:
Large Clearance Pestle
Small Clearance Pestle
Unit Size:
Each
Description
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Capacity (English) | 1.35 oz. |
| Capacity (Metric) | 40 mL |
| Product Type | Small Clearance Pestle |
| For Use With (Application) | To form final homogenate |
| For Use With (Equipment) | Dounce all Glass tissue grinders |
| Capacity | 1.35 oz., 40mL |