Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn™ Economy Columns
Product Code. 10647955
Change view
Click to view available options
Length (Metric):
40 mm
100 mm
150 mm
300 mm
500 mm
Column Capacity:
1.5 mL
4 mL
6 mL
12 mL
19 mL
Quantity:
1
10
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Column Capacity Quantity unitSize
10647955 150 mm 6 mL 1 Each
15244095 100 mm 4 mL 1 Each
12746768 300 mm 12 mL 1 Each
12756768 500 mm 19 mL 1 Each
11702354 40 mm 1.5 mL 10 Pack of 10
11732354 300 mm 12 mL 10 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10647955 Supplier DWK Life Sciences Supplier No. 4204010715
Only null left
Add to basket
Add to basket
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
Use columns with ion-exchange, affinity, gel filtration, and hydrophobic interaction media.
  • Durable borosilicate glass
  • Precision-ground column ends form positive seals with endcaps so they don't leak or “creep” under pressure
  • Threaded, easy to remove reservoir cap won't pop off under pressure
  • Reservoir accepts accessory funnels and glass reservoirs
  • Luer-Lock polypropylene inlet and outlet fittings
  • Bed support has a 20μm polyethylene disc that retains most media for low-pressure aqueous chromatography
  • Columns accept flow adapters for adjusting Flex-Column bed height (order adapters separately)
Flow adapters not available
TRUSTED_SUSTAINABILITY
Specifications
Endcapped Yes
For Use With (Application) Ion exchange, affinity, gel filtration and hydrophobic interaction media
Quantity 1
Temperature (Metric) Range 0 to 50°C
Diameter (Metric) Inner 7 mm
Type Economy Chromatography Column
Max. Pressure 14.5 psi
Pore Size 20 μm
Column Capacity 6 mL
Length (Metric) 150 mm
Material Borosilicate Glass Type I Class A
Support Material HDPE
Connector Types Male luer lock
Cross Sectional Area 0.39cm3
Sterilization Method 2N NaOH or 100% Ethanol