Learn More
DWK Life Sciences Kimble™ Kontes™ Incubation Flask Stoppers and Center Well
Rubber stopper for flask sidearm provides a secure double seal
Brand: DWK Life Sciences 882311-0000
Additional Details : Weight : 0.80000kg
Description
Sidearm Flask Stoppers
- Rubber stopper for flask sidearm provides a secure double seal
- Sidearm stopper can be punctured with a syringe needle
- Stoppers fit 10 and 25mL flasks
Order Info
Top and sidearm stoppers are not interchangeable.
Specifications
Rubber | |
Off-center hole | |
144 | |
Sleeved stopper |