Learn More
DWK Life Sciences Kimble™ Kontes™ Kuderna-Danish Evaporative Concentrator: Flask Only
Product Code. 10328454
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10328454 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10328454 Supplier DWK Life Sciences Supplier No. 5700010250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
For evaporative concentration
  • 24/40 top joint
  • 19/22 bottom joint
Specifications
For Use With (Equipment) Evaporative Concentrator
Product Type Extraction Flask
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Includes Flask
Standard Taper Joint Bottom 19/22
Standard Taper Joint Top 24/40