Learn More
DWK Life Sciences Kimble™ Kontes™ Potter-Elvehjem Tissue Grinder
Click to view available options
Product Type:
Tissue Grinder
Tissue Grinder Tube
Unit Size:
Each
Description
- Radial serrations on the lower portion of the pestle deliver the homogenate into the cylindrical portion of the tube
- Tube has a convenient pouring lip
- Stainless-steel pestle rod
- Clearance between pestles and tubes is 0.004 to 0.006 in.
- Working Capacity: 17mL
Specifications
Specifications
| Size | 23 |
| Clearance (English) | 0.004 to 0.006 in. |
| Clearance (Metric) | 0.00015 to 0.0002mm |
| Capacity (English) | 3.38 oz. |
| Capacity (Metric) | 17 mL |
| Material | Stainless Steel |
| Product Type | Tissue Grinder Tube |
| For Use With (Equipment) | Potter-Elvehjem tissue grinder |