Learn More
DWK Life Sciences Kimble™ Kontes™ Rectangular TLC Developing Tanks
Product Code. 11738482
Change view
Click to view available options
Closure Type:
Latch Lid
Nonslip
Standard
Depth (English):
10.47 in.
10.75 in.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type Depth (English) Unit Size
11738482 Standard 10.75 in. Each
12766728 Nonslip 10.47 in. Each
12776728 Latch Lid 10.75 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11738482 Supplier DWK Life Sciences Supplier No. 4161800000
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Molded glass tanks for development of any size TLC plate up to 20 x 20cm
Molded glass tanks for development of any size TLC plate up to 20 x 20cm
  • Tanks: K416180-0000, K416185-0000, K416190-0000
  • Lids: K416181-0000, K416186-0000
Specifications
Closure Type Standard
Material Soda Lime Glass
Type Developing Tank
Holds 20 x 20 cm or four 5 x 20 cm Plates
Shape Rectangular
Dimensions (L x W) 4 x 11.75 in.
Length (English) 4 in.
Length (Metric) 10.16 cm
Width (English) 11.75 in.
Width (Metric) 29.8 cm
Depth (English) 10.75 in.
Depth (Metric) 27.3 cm