Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Tube For All-Glass Dounce Tissue Grinders
Product Code. 10432385
Change view
Click to view available options
Capacity:
0.06 oz., 2mL
0.23 oz., 7mL
0.5 oz., 15mL
1.35 oz., 40mL
3.38 oz., 100mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10432385 0.23 oz., 7mL Each
10724413 3.38 oz., 100mL Each
11816271 0.5 oz., 15mL Each
11864361 1.35 oz., 40mL Each
11883722 0.06 oz., 2mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10432385 Supplier DWK Life Sciences Supplier No. 8853030007
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Quantity Case of 1
Capacity (English) 0.23 oz.
Capacity (Metric) 7 mL
Product Type Tissue Grinder Tube
For Use With (Equipment) Dounce all Glass tissue grinders
Capacity 0.23 oz., 7mL