Learn More
DWK Life Sciences Kimble™ Kontes™ Tube For Potter-Elvehjem Tissue Grinder
Description
- Graduations are fused onto the glass
- Clearance between pestle and tube
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Size | 19 |
| Product Type | Tissue Grinder Tube |
| For Use With (Equipment) | Potter-Elvehjem tissue grinder |