Learn More
DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 15 mL Capacity
Click to view available options
Disposable:
Reusable
Single-use
Length (Metric):
118 mm
126 mm
130 mm
Unit Size:
Pack of 12
Pack of 125
Graduations:
0.1 mL
Permanent Red, 0.1 mL
Description
Withstand centrifugation up to 2980 RCF
- With conical shaped bottom
- Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
Specifications
Specifications
| Certifications/Compliance | USP, ASTM E 438 |
| Capacity (Metric) | 15 mL |
| Disposable | Single-use |
| Material | Borosilicate Glass |
| Shape | Conical |
| Length (Metric) | 126 mm |
| Bottom Shape | Conical |
| Closure Type | Not Included |
| Diameter (Metric) | 17 mm |
| Graduated | No |
| Show More |