Learn More
DWK Life Sciences™ Kimble™ Kontes™ Distillation Apparatus for Acid-Soluble, Acid-Insoluble, and Extractable Sulfides
Product Code. 10103762
Change view
Click to view available options
Type:
Fritted Gas Dispersion Tube
Height (English):
9.84 in.
Height (Metric):
250mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Height (English) Height (Metric) Unit Size
10103762 Fritted Gas Dispersion Tube 9.84 in. 250mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10103762 Supplier DWK Life Sciences™ Supplier No. 9565000023
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Compliance
Meets criteria for U.S. EPA Methods 9030A and 9031 for SW-846
Specifications
Includes Hollow fritted cylinder at the end of the tube
Height (Metric) 250mm
Type Fritted Gas Dispersion Tube
Material Borosilicate Glass
Height (English) 9.84 in.
Certifications/Compliance USP Type I and ASTM E438, Type I, Class A
For Use With (Equipment) Distillation apparatus
Diameter (Metric) Outside 13mm
Diameter (English) Outside 0.51 in.