Learn More
DWK Life Sciences Kimble™ Kontes™ Incubation Flasks
For metabolic studies of biological materials
Brand: DWK Life Sciences 8823600010
Additional Details : Weight : 0.80000kg
Description
Ideal for measuring C14O2 yields following incubation, in vitro incubations, insulin bioassays, persulfate oxidations, and distillation of any volatile acid or base. Also suitable for use with phosphate or bicarbonate buffers when acid or alkali is introduced at the end of a reaction.
- Available with or without sidearm
- Flask with sidearm enables syringe needle entry without disturbing established conditions
Order Info
Stoppers and center well sold separately.
Specifications
Rubber | |
Borosilicate Glass | |
2 in. | |
Sidearm | |
7 cm | |
3.1 cm |
10 mL | |
For measuring C14O2 yields following incubation, in-vitro incubations, Insulin bioassays, Persulfate oxidations and distillation of any volatile Acid or base | |
5 cm | |
2.7 in. | |
1.2 in. | |
Stopper |