Learn More
DWK Life Sciences Kimble™ Kontes™ Pestle For All-Glass DUALL™ Tissue Grinders
Grinding efficiency is greatly improved when this tube is used. Available in sizes 20 - 23.
Brand:  DWK Life Sciences 885451-0021
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10102062
  • € 72.30 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
Grinding efficiency is greatly improved when this tube is used. Available in sizes 20 - 23.
  • Combines both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate.
  • This ground glass model is ideal for connective tissue such as muscle, heart and lung
  • Construction is strong enough to allow the pestle to be motor-driven
  • Clearance between pestles and tubes is 0.004 in. to 0.006 in.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
21
Tissue grinder pestle
Duall™ all Glass tissue grinders