missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint

Product Code. 11848462
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
250 mL
Diameter (Metric) Outer:
4 cm
5 cm
8.5 cm
Standard Taper Joint:
19/22
24/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Standard Taper Joint unitSize
11848462 250 mL 8.5 cm 24/40 Each
11773143 50 mL 5 cm 19/22 Each
12698341 25 mL 4 cm 19/22 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11848462 Supplier DWK Life Sciences Supplier No. 4225300250
 Restricted Item
Estimated Shipment: 23-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy

Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids

  • Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
  • Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
  • Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
  • Adjust spray pattern by varying the position of thumb on vent hole
  • Autoclavable (after cap, loosening ring, and O-ring are removed)

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 8.45 oz.
Capacity (Metric) 250 mL
Connections Screw thread ground joint, connecting cap, loosening ring
Material Glass
Quantity 1 Case
Type TLC Reagent Sprayer
For Use With (Application) To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography
For Use With (Equipment) Standard TLC Reagents and Most Corrosive Liquids
Operating Pressure 3 to 5 psi
Standard Taper Joint 24/40
Height (English) 9.01 in.
Height (Metric) 22.9 cm
Diameter (English) Outer 3.34 in.
Diameter (Metric) Outer 8.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.