Learn More
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint
Product Code. 12698341
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
250 mL
Diameter (Metric) Outer:
4 cm
5 cm
8.5 cm
Standard Taper Joint:
19/22
24/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Standard Taper Joint Unit Size
12698341 25 mL 4 cm 19/22 Each
11848462 250 mL 8.5 cm 24/40 Each
11773143 50 mL 5 cm 19/22 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12698341 Supplier DWK Life Sciences Supplier No. 4225300025
  • ONLINE EXCLUSIVE PRICE
    € 348.60
    € 203.78 / Pack of 1
    Save € 144.82
    41% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
  • Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
  • Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
  • Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
  • Adjust spray pattern by varying the position of thumb on vent hole
  • Autoclavable (after cap, loosening ring, and O-ring are removed)
Specifications
Autoclavable Autoclavable
Capacity (English) 0.84 oz.
Capacity (Metric) 25 mL
Connections Screw thread ground joint, connecting cap, loosening ring
Material Glass
Quantity 1 Case
Type TLC Reagent Sprayer
For Use With (Application) To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography
For Use With (Equipment) Standard TLC Reagents and Most Corrosive Liquids
Operating Pressure 3 to 5 psi
Standard Taper Joint 19/22
Height (English) 5.78 in.
Height (Metric) 14.7 cm
Diameter (English) Outer 1.57 in.
Diameter (Metric) Outer 4 cm