Learn More
DWK Life Sciences Kimble™ Screw Caps for Disposable Glass Tubes
Linerless, one-piece closures with inner sealing ring
€ 223.00
Specifications
Product Type | Screw Cap |
---|---|
Certifications/Compliance | ASTM E982 |
Autoclavable | Autoclavable |
Material | Polypropylene |
Description
Manufactured from polypropylene. Kimble Chase™ Screw Caps for Disposable Glass Tubes are autoclavable.
- Polypropylene
- Autoclavable
Compliance
Designed from ASTM™ E 982
Specifications
Screw Cap | |
ASTM E982 | |
Autoclavable | |
Polypropylene |