missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Straight-Sided Serum Stoppers
Click to view available options
Material:
Chlorobutyl Rubber, PTFE
Diameter (Metric) Outer:
13 mm
20 mm
Unit Size:
Pack of 100
Beschreibung
- For use with 13 and 20mm serum-finished bottles and vials
- Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
- Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors or natural rubber latex
- Applicable to aqueous solutions with a pH range of 2 to 10
- PTFE facing improves chemical compatibility and minimizes surface interactions
Bestellinformation
Durometer 50.
Spezifikation
Spezifikation
| Description | Latex free |
| For Use With (Application) | For research and pharmaceutical packaging applications |
| For Use With (Equipment) | 13 mm GPI Aluminum Seal |
| Hardness | 50 Durometer |
| Color | Gray |
| Diameter (Metric) Outer | 13 mm |
| Material | Chlorobutyl Rubber, PTFE |
| Product Type | Plug Stopper |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur