Learn More
DWK Life Sciences Kimble™ Straight-Sided Serum Stoppers
Product Code. p-78890
Click to view available options
Material:
Chlorobutyl Rubber, PTFE
Diameter (Metric) Outer:
13 mm
20 mm
Unit Size:
Pack of 100
Product Code. 10632174
Brand: DWK Life Sciences 73811T21
Only null left
Add to basket
Add to basket
For research and pharmaceutical packaging applications
  • For use with 13 and 20mm serum-finished bottles and vials
  • Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
  • Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors or natural rubber latex
  • Applicable to aqueous solutions with a pH range of 2 to 10
  • PTFE facing improves chemical compatibility and minimizes surface interactions
Order Info
Durometer 50.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP, EP, JP
Description Latex free
For Use With (Application) For research and pharmaceutical packaging applications
For Use With (Equipment) 20 mm GPI Aluminum Seal
Hardness 50 Durometer
Color Gray
Diameter (Metric) Outer 20 mm
Material Chlorobutyl Rubber, PTFE
Product Type Plug Stopper