Learn More
Kimberly-Clark™ Kimcare Industrie™ Diabolo Hand Cleanser
Description
- For general purpose hand cleansing
- Cleans and cares for working hands
- Balanced pH formula with emollients
Specifications
Specifications
| Product Type | Hand Cleanser |
| Color | Orange |
| Dimensions (L x W x H) | 24.4 x 17.23 x 13.2 cm |
| For Use With (Equipment) | Kimberly-Clark Professional hand cleanser dispenser |
| Length (English) | 10.9 in. |
| Length (Metric) | 27.8 cm |
| Quantity | 2 Pk. |
| Volume (English) | 118.3 oz. |
| Volume (Metric) | 3.5 L |