Learn More
Kimtech™ M3 Certified Sterile Pleat-Style Face Mask with Ties
GREENER_CHOICE
Product Code. 10702121
Click to view available options
Includes:
18 cm (7 in.) Ties
Unit Size:
Pack of 20
Product Code. 10702121
Brand: Kimtech™ 62467
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sterile pleat-style masks with high bacterial and particle filtration, suitable for EU GMP ISO 5 Class A sterile cleanrooms. Latex- and silicone-free Kimtech™ M3 Certified Mask reduces contamination with a low-lint polyethylene film outer layer and Bicosof™ inner fabric.
  • Bacterial and particle filtration to avoid contamination
  • Suitable for use in EU GMP ISO 5 Class A sterile cleanrooms environments and laboratories, and provided with supporting technical information to demonstrate regulatory compliance
  • Available as white, sterile face masks and with intuitive and comfortable ties
  • Constructed from a low-lint polyethylene film outer layer and secure Bicosof™ inner fabric for enhanced protection
  • Contain no natural rubber latex or silicone, reducing the risks of skin irritation for the wearer
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material Polyethylene
Size Standard
Sterility Sterile
Style Pleated
Certifications/Compliance ISO 14001, ISO 9001, CE Simple, Industrial Use Only
Cleanroom Class Class 1 (ISO 3)
Disposable Single-use
Filter Efficiency 3.0 micron
Includes 18 cm (7 in.) Ties
Latex Free Yes
Model 62467
Pore Size 0.1 μm (PFE), 3 μm (BFE)
Series Kimtech™ M3 Sterile