Learn More
Invitrogen™ KIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579557
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human MCF-7 whole cell, human 293T whole cell, human HepG2 whole cell, rat RH35 whole cell, rat NRK whole cell, mouse HEPA1-6 whole cell, mouse NIH/3T3 whole cell. IHC: human oesophagus squama cancer tissue.
Kin is a nuclear protein that forms intranuclear foci during proliferation and is redistributed in the nucleoplasm during the cell cycle. Short-wave ultraviolet light provokes the relocalization of the protein, suggesting its participation in the cellular response to DNA damage. Originally selected based on protein-binding with RecA antibodies, the mouse protein presents a limited similarity with a functional domain of the bacterial RecA protein, a characteristic shared by the human ortholog.
Specifications
| KIN | |
| Polyclonal | |
| Unconjugated | |
| Kin | |
| antigenic determinant of rec-A protein; antigenic determinant of rec-A protein homolog; binding to curved DNA; Btcd; DNA/RNA-binding protein KIN17; HsKin17 protein; KIN; KIN, antigenic determinant of recA protein; KIN, antigenic determinant of recA protein homolog; KIN17; Kin17 DNA and RNA binding protein; Rts2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16588, 22944, 689197 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.01mg sodium azide | |
| O60870, Q8K339 | |
| Kin | |
| E.coli-derived human KIN recombinant protein (Position: K3-K202). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.