missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kindlin Antibody (4A5.14), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08647APC
This item is not returnable.
View return policy
Description
Kindlin Monoclonal antibody specifically detects Kindlin in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Kindlin | |
| Monoclonal | |
| APC | |
| C20orf42fermitin family homolog 1, chromosome 20 open reading frame 42, fermitin family homolog 1 (Drosophila), fermitin family member 1, FLJ20116, KIND1DTGCU2, kindlerin, kindlin 1, Kindlin syndrome protein, Kindlin-1, UNC112 related protein 1, UNC112A, Unc-112-related protein 1, URP1FLJ23423 | |
| Recombinant protein containing Kindlin (residues 216-677) (Uniprot: Q9BQL6) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 4A5.14 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 55612 | |
| Store at 4°C in the dark. | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction