missing translation for 'onlineSavingsMsg'
Learn More
Learn More
kinectin Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32261AF750
This item is not returnable.
View return policy
Description
kinectin Polyclonal antibody specifically detects kinectin in Human samples. It is validated for Western Blot
Specifications
| kinectin | |
| Polyclonal | |
| Western Blot | |
| kinectin, kinectin 1 (kinesin receptor), MGC133337 | |
| The immunogen this antibody was made to, maps to a region between residue 150 to 200 of human Kinectin using the numbering given in entry NP_001072989.1 (GeneID 3895). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 3895 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction