Learn More
HMC Europe Vertical Autoclave Air Filters
Exhaust air filter. Safety for individuals and the environment. Option for safe, emission-free sterilization of highly infectious germs and genetically modified materials. Not for applications requiring FDA approval.
€ 235.70 - € 3703.06
Specifications
For Use With (Application) Sterilization of exhaust air
View More Specs
Products 3
Product Code Brand Product Type For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Product Type For Use With (Equipment) Price Quantity & Availability  
10592384
View Documents Promotion Details
HMC Europe
51011
Exhaust Air Filter HMC-HG autoclave
€ 3651.06
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
12672925
View Documents Promotion Details
HMC Europe
50019A
Replacement Air Filter HMC-HV autoclave
€ 235.70
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-10-2024
to see stock.
Add to basket
10221832
View Documents Promotion Details
HMC Europe
50004
Exhaust Air Filter HMC-HV autoclave
€ 3703.06
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-10-2024
to see stock.
Add to basket
Description
Description
During the venting phase, the steam displaces unsterilized residual air from the chamber and passes it through a filter system. The surrounding air is protected against contamination as the cycle is running. Naturally, in-line sterilization of the exhaust air filter is performed. Due to the design, condensate sterilization is a matter of course. Any condensate which precipitates flows back into the chamber and gets sterilized. Discharge can be done safely.
Specifications
Specifications
Sterilization of exhaust air