Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand: HMC Europe 51009
Additional Details : Weight : 0.50000kg
Description
For use with the HMC-HG and HMC-HV autoclaves.Specifications
345 mm | |
305 mm | |
33.8 cm | |
80 L (Total), 50 L (Nominal) | |
9 in. | |
Wire mesh basket | |
338 x 235 mm |
Wire mesh basket | |
13.3 in. | |
13.2/21.13 gal. | |
HG 50/80 autoclave | |
235 mm | |
Stainless Steel |