Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand: HMC Europe 50007
Additional Details : Weight : 0.50000kg
Description
For sue with the HMC-HG and HMC-HV autoclaves
Specifications
11 in. | |
13.2 gal. | |
HV 50 autoclave | |
22.5 cm | |
Stainless Steel |
28 cm | |
50 L | |
8.85 in. | |
Stackable basket | |
280 x 225 mm |