Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand:  HMC Europe 50028
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10318092
  • € 582.32 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
For use with the HMC-HG and HMC-HV autoclaves.
Specifications
Specifications
11 in.
13.2 gal.
HV 50 autoclave
225 mm
Stainless Steel
280 mm
50 L
8.85 in.
With collecting pan
280 x 225 mm