Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand: HMC Europe 51019
Additional Details : Weight : 0.99000kg
Description
For use with the HMC-HG and HMC-HV autoclaves.Specifications
315 mm | |
345 mm | |
80 L (Total), 50 L (Nominal) | |
12.2 in. | |
Wire mesh basket | |
345 x 310 mm |
13.3 in. | |
13.2/21.13 gal. | |
HG 80 autoclave | |
310 mm | |
Stainless Steel |